Merck
  • Activation of cAMP and mitogen responsive genes relies on a common nuclear factor.
Activation of cAMP and mitogen responsive genes relies on a common nuclear factor.
Nature (1994-07-21)
J Arias, A S Alberts, P Brindle, F X Claret, T Smeal, M Karin, J Feramisco, M Montminy
ABSTRACT
A number of signalling pathways stimulate transcription of target genes through nuclear factors whose activities are primarily regulated by phosphorylation. Cyclic AMP regulates the expression of numerous genes, for example, through the protein kinase-A (PKA)-mediated phosphorylation of transcription factor CREB at Ser 133. Although phosphorylation may stimulate transcriptional activators by modulating their nuclear transport or DNA-binding affinity, CREB belongs to a class of proteins whose phosphorylation appears specifically to enhance their trans-activation potential. Recent work describing a phospho-CREB binding protein (CBP) which interacts specifically with the CREB trans-activation domain prompted us to examine whether CBP is necessary for cAMP regulated transcription. We report here that microinjection of an anti-CBP antiserum into fibroblasts can inhibit transcription from a cAMP responsive promoter. Surprisingly, CBP also cooperates with upstream activators such as c-Jun, which are involved in mitogen responsive transcription. We propose that CBP is recruited to the promoter through interaction with certain phosphorylated factors, and that CBP may thus play a critical role in the transmission of inductive signals from cell surface receptor to the transcriptional apparatus.
Sigma-Aldrich
KAT3A (518-1207), GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
CBP (1319-1710), GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.