
Orphenadrine Related Compound E

Synonym(s):
N,N-Dimethyl-2-[(3-methylphenyl)phenylmethoxy]ethanamine, N,N-Dimethyl-2-[phenyl(m-tolyl)methoxy]ethanamine
Empirical Formula (Hill Notation):
C18H23NO
CAS Number:
Molecular Weight:
269.38
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2