
Fraxetin

Synonym(s):
7,8-Dihydroxy-6-methoxycoumarin, Fraxetin
Empirical Formula (Hill Notation):
C10H8O5
CAS Number:
Molecular Weight:
208.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing