
Dirhodium(II)tetrakisoctanoate

Synonym(s):
Rhodium(II) octanoate, dimer
Linear Formula:
[[CH3(CH2)6CO2]2Rh]2
CAS Number:
Molecular Weight:
778.63
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing