Skip to Content
Merck
Dimethylamine-13C2,15N hydrochloride 99 atom % 13C, 98 atom % 15N

Dimethylamine-13C2,15N hydrochloride

Dimethylamine-13C2,15N hydrochloride 99 atom % 13C, 98 atom % 15N
Linear Formula:
(13CH3)215NH ·HCl
CAS Number:
Molecular Weight:
84.52

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing