
Anti-Bovine Albumin antibody produced in rabbit

Synonym(s):
Anti-Albumin, bovine serum (BSA)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3