![5-Bromo-5-nitro-1,3-dioxane analytical standard](/deepweb/assets/sigmaaldrich/product/structures/331/746/1afc6621-1ae6-467a-a942-914afbe9f71d/640/1afc6621-1ae6-467a-a942-914afbe9f71d.png)
5-Bromo-5-nitro-1,3-dioxane
![5-Bromo-5-nitro-1,3-dioxane analytical standard](/deepweb/assets/sigmaaldrich/product/structures/331/746/1afc6621-1ae6-467a-a942-914afbe9f71d/640/1afc6621-1ae6-467a-a942-914afbe9f71d.png)
Empirical Formula (Hill Notation):
C4H6BrNO4
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing