
3-Methoxythiophenol

Synonym(s):
3-Mercaptoanisole, 3-Methoxybenzenethiol
Linear Formula:
CH3OC6H4SH
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing