Merck
  • Involvement of intracellular transport in TREK-1c current run-up in 293T cells.
Involvement of intracellular transport in TREK-1c current run-up in 293T cells.
Channels (Austin, Tex.) (2017-01-14)
Naaz Andharia, Ancy Joseph, Mikio Hayashi, Masayoshi Okada, Hiroko Matsuda
ABSTRACT
The TREK-1 channel, the TWIK-1-related potassium (K
Sigma-Aldrich
Anti-TREK-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.