Merck
  • Magnetic porous β-cyclodextrin polymer for magnetic solid-phase extraction of microcystins from environmental water samples.
Magnetic porous β-cyclodextrin polymer for magnetic solid-phase extraction of microcystins from environmental water samples.
Journal of chromatography. A (2017-05-14)
Wenmin Zhang, Mingxia Lin, Meili Wang, Ping Tong, Qiaomei Lu, Lan Zhang
ABSTRACT
Microcystins (MCs) are cyclic heptapeptide toxins and tumor promoters produced by cyanobacteria, which threaten the health of humans. In this study, magnetic porous β-cyclodextrin polymer (Fe