Merck
  • Curcumin reverses benzidine-induced epithelial-mesenchymal transition via suppression of ERK5/AP-1 in SV-40 immortalized human urothelial cells.
Curcumin reverses benzidine-induced epithelial-mesenchymal transition via suppression of ERK5/AP-1 in SV-40 immortalized human urothelial cells.
International journal of oncology (2017-03-06)
Zhiqi Liu, Jie Liu, Li Zhao, Hao Geng, Jiaxing Ma, Zhiqiang Zhang, Dexin Yu, Caiyun Zhong
ABSTRACT
Overexposure to benzidine has been manifested as an important cause of bladder cancer. However, the molecular mechanism of benzidine-induced malignancy is still insufficiently interpreted. Epithelial-mesenchymal transition (EMT) is a crucial pathophysiological process in embryonic development as well as initiation and development of epithelium-originated malignant tumors. The role of extracellular regulated protein kinase 5 (ERK5) in benzidine-meditated bladder cancer development has not been explored. In the present study, we explored the role of ERK5/AP-1 pathway in benzidine-induced EMT in human normal urothelial cells and the intervention effect of curcumin on bezidine-induced EMT. We found that benzidine-induced EMT in SV-40 immortalized human urothelial cells (SV-HUC-1) at low concentrations. We detected that ERK5/AP-1 pathway was notably activated. Specific ERK5 inhibitor, XMD8-92 was applied to determine the role of ERK5 in benzidine-induced EMT. Results indicated that XMD8-92 reversed the EMT process. Furthermore, curcumin effectively attenuated benzidine-induced urocystic EMT by suppressing ERK5/AP-1 pathway. In conclusion, the present study revealed the positive role of ERK5/AP-1 in benzidine-provoked urocystic EMT and the curcumin promising use in bladder cancer prevention and intervention via ERK5/AP-1 pathway.
Sigma-Aldrich
XMD8-92, ≥98% (HPLC)