Merck
  • Effect of SP600125 on the mitotic spindle in HeLa Cells, leading to mitotic arrest, endoreduplication and apoptosis.
Effect of SP600125 on the mitotic spindle in HeLa Cells, leading to mitotic arrest, endoreduplication and apoptosis.
Molecular cytogenetics (2016-12-08)
Donia Mili, Kaouthar Abid, Imed Rjiba, Abderraouf Kenani
ABSTRACT
The JNK inhibitor SP600125 strongly inhibits cell proliferation in many human cancer cells by blocking mitosis progression and inducing cell death. Despite, all this study, the mechanism by which SP600125 inhibits mitosis-related effects in human cervical cells (HeLa cells) remains unclear. In this study, we investigated the effects of SP600125 on the cell viability, cell cycle, and on the spindle assembly during mitosis in HeLa cells. To explore this approach, we used a viability test, an immunofluorescence microscopy to detect Histone phosphorylation and mitotic spindle aberrations. Apoptosis was characterised using Western Blotting. Treatment of HeLa cells with varying concentrations of SP600125 induces significant G2/M cell cycle arrest with elevated phosphorylation of histone H3 within 48 h, and endoreduplication after 48 h. SP600125 also induces significant abnormal mitotic spindle. High concentrations of SP600125 (20 μM) induce disturbing microtubule assembly in vitro. Additionally, SP600125- induced delayed apoptosis and cell death was accompanied by significant poly ADP-ribose polymerase (PARP) cleavage and caspase-3 activation in the late phase (at 72 h). Our results confirmed that SP600125 induce mitosis arrest in G2/M, endoreduplication, mitotic spindle aberrations and apoptosis.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Atto 488 antibody produced in goat, ~1 mg/mL
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Pricing and availability is not currently available.