Merck
  • Application of a novel 1,1'-dimethylferricinium dye for the determination of uric acid in urine.
Application of a novel 1,1'-dimethylferricinium dye for the determination of uric acid in urine.
Applied biochemistry and biotechnology (1994-01-01)
K B Male, J H Luong, M Trani
ABSTRACT
Water-soluble 2-hydroxypropyl-beta-cyclodextrin (Hp-beta-CyD), a cyclic and nonreducing oligosaccharide was used to enclose a hydrophobic guest molecule 1,1'-dimethylferrocene (DMF) to form a water-soluble yellow complex. At high concentrations (300 mM), Hp-beta-CyD enclosed up to 100 mM DMF. The yellow complex was electrochemically oxidized (platinum vs Ag/AgCl poised at +450 mV) to form a blue dye, 1,1'-dimethylferricinium (DMF+). This is a one-electron transfer process and the ferricinium cation formed exhibited an absorption peak at 650 nm. The concentrated DMF+ was stable for at least 4 mo at 4 degrees C and insensitive to a wide pH variation (pH 2-11). Application of the novel DMF+ complex as a colorimetric dye for the determination of uric acid in urine was successfully demonstrated. The reaction between the dye and uric acid is almost instantaneous and decrease in absorbance caused by the reduction of 1,1'-dimethylferricinium to 1,1'-dimethylferrocene can be followed at 650 nm. The results obtained agreed well with those of the reference reversed-phase HPLC method.
Sigma-Aldrich
1,1′-Dimethylferrocene, 95%