Merck
  • Methylation and restriction endonuclease cleavage of linear Z-DNA in the presence of hexamminecobalt (III) ions.
Methylation and restriction endonuclease cleavage of linear Z-DNA in the presence of hexamminecobalt (III) ions.
Nucleic acids research (1986-09-25)
G Soslau, J Parker, J W Nelson
ABSTRACT
These studies employed the synthetic linear DNA, poly dGdC, in the B and cobalt hexammine chloride (Co)-induced Z form to determine the effect of conformation on protein-DNA interactions. The rate of the reaction of the restriction endonucleases, Hha I and Cfo I, are reduced with Z DNA as compared to B DNA. The ability of both restriction endonucleases to react with an aggregate form of Z DNA (Z* DNA) is found to depend upon how the Z* DNA is formed. When Z* DNA is induced by low concentrations of Co (50 microM), the endonucleases remain active. In the presence of 100 microM Co, which causes increased aggregation, the endonucleases are inactive. The Hha I DNA methyltransferase reacts at equal rates with the B, Z and low cobalt Z* forms and at a greatly reduced rate with the high cobalt Z* form. These results are significantly different than those observed with Z form dGdC tracts inserted into circular DNA molecules.
Sigma-Aldrich
Hexaamminecobalt(III) chloride, 99%
Sigma-Aldrich
Hexammine cobalt(III) chloride, for use in transformations, X-ray crystallography and NMR