Merck
  • Cs4Mo5P2O22: a first Strandberg-type POM with 1D straight chains of polymerized [Mo5P2O23]6- units and moderate second harmonic generation response.
Cs4Mo5P2O22: a first Strandberg-type POM with 1D straight chains of polymerized [Mo5P2O23]6- units and moderate second harmonic generation response.
Chemical communications (Cambridge, England) (2012-11-28)
Ying Wang, Shilie Pan, Hongwei Yu, Xin Su, Min Zhang, Fangfang Zhang, Jian Han
ABSTRACT
A noncentrosymmetric chiral Strandberg-type polyoxometalate based material, Cs(4)Mo(5)P(2)O(22), that contains unusual one dimensional chains constructed from [Mo(5)P(2)O(23)](6-) units has been reported. Cs(4)Mo(5)P(2)O(22) shows a moderate SHG response, and is phase-matchable.
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis
Pricing and availability is not currently available.