Merck
  • Hyperammonaemia due to cobalamin malabsorption in a cat with exocrine pancreatic insufficiency.
Hyperammonaemia due to cobalamin malabsorption in a cat with exocrine pancreatic insufficiency.
Journal of feline medicine and surgery (2012-08-22)
Takayuki Watanabe, Katsuichiro Hoshi, Chunhua Zhang, Yuzuru Ishida, Ikuo Sakata
ABSTRACT
A 10-year-old domestic shorthair cat showed anorexia, lethargy and ptyalism with hyperammonaemia. Portosystemic shunts were not identified by computed tomography angiography. Biopsy results revealed mild interstinal nephritis and no lesion in the liver. Analysis of urine revealed the presence of a high methylmalonic acid (MMA) concentration. Serum cobalamin (vitamin B(12)) and serum feline trypsin-like immunoreactivity levels were also markedly low. The cat was diagnosed as having exocrine pancreatic insufficiency (EPI). After 5 weeks of parenteral cobalamin supplementation, serum cobalamin concentration had increased and urinary MMA concentration had decreased. This case suggests that hyperammonaemia may be caused by accumulation of MMA due to cobalamin malabsorption secondary to feline EPI.
Product Description
Supelco
Methylmalonic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Sigma-Aldrich
Methylmalonic acid, 99%
Pricing and availability is not currently available.