Merck
  • Ionic liquid acceleration of solid-phase suzuki-miyaura cross-coupling reactions.
Ionic liquid acceleration of solid-phase suzuki-miyaura cross-coupling reactions.
Organic letters (2002-08-31)
Jefferson D Revell, A Ganesan
ABSTRACT
[reaction: see text] Room-temperature ionic liquids promote various transition metal-catalyzed reactions in the solution phase. Here, for the first time, we show that these effects are translatable to solid-phase reactions. The Suzuki-Miyaura cross-coupling of 4-iodophenol immobilized on polystyrene-Wang resin with various arylboronic acids was significantly accelerated by the ionic liquid 1-butyl-3-methylimidazolium tetrafluoroborate ([bmim][BF(4)(-)]).
Sigma-Aldrich
4-Iodophenol, 99%
Pricing and availability is not currently available.