Merck
  • 2-Amino-7-deazaadenine forms stable base pairs with cytosine and thymine.
2-Amino-7-deazaadenine forms stable base pairs with cytosine and thymine.
Bioorganic & medicinal chemistry letters (2001-12-12)
Akimitsu Okamoto, Kazuo Tanaka, Isao Saito
ABSTRACT
2-Amino-7-deazaadenine ((AD)A) was incorporated into oligodeoxynucleotides (ODN) and their base-pairing properties with natural nucleobases were investigated. In melting temperature (T(m)) experiments, the duplex containing an (AD)A/C base pair showed a high stability comparable to that containing (AD)A/T base pair. Destabilization of the duplex usually observed for existing degenerate bases was not observed. However, the incorporation efficiency of dCTP was only 1.8% for TTP in single-nucleotide insertion reactions using polymerase.
Sigma-Aldrich
6-Amino-7-deazapurine, 97%