Merck
  • [QacEdelta1 gene frequency and biocide resistance in extended-spectrum beta-lactamase producing enterobacteriaceae clinical isolates].
[QacEdelta1 gene frequency and biocide resistance in extended-spectrum beta-lactamase producing enterobacteriaceae clinical isolates].
Revista de investigacion clinica; organo del Hospital de Enfermedades de la Nutricion (2013-03-22)
Josefina Pastrana-Carrasco, Jesús Ulises Garza-Ramos, Humberto Barrios, Rayo Morfin-Otero, Eduardo Rodríguez-Noriega, Juan Manuel Barajas, Sandra Suárez, Rita Díaz, Guadalupe Miranda, Fortino Solórzano, Jesús Contreras, Jesús Silva-Sánchez
ABSTRACT
To determine the frequency of the gene qacEdelta1 and characterize the resistance to biocides of extended-spectrum beta-lactamases producing enterobacteriaceae (ESBL-PE) obtained from clinical isolates causing nosocomial infections. In total 59 ESBL-PE causing nosocomial infections were included: Klebsiella pneumoniae (35) and Enterobacter cloacae (24). Minimal inhibitory concentration (MIC) was tested for chlorhexidine (CHX) and benzalkonium chloride (CLBZ) by agar dilution technique. Amplification of the SHV, TLA-1 and qacEdelta1 genes were performed by PCR using specific primers and plasmid identification was done by alkaline lysis method. Matting experiments were obtained on solid agar method. Chlorhexidine-resistance was found in 100% of the ESBL-PE and benzalkonium chloride-resistance in 80%. In 68% of the biocides-resistant strains the qacEdelta1 gene was present. The 66% of resulting transconjugants were resistant to CHX and the gene qacEdelta1 was detected in 55%. The qacEdelta1 gene of antiseptic resistance is widespread in the EP-ESBL and can be transferred horizontally. Thus it is advisable to use combinations of antiseptics, as recommended in the literature, to avoid selection of multiresistant bacteria in hospitals, causing nosocomial infections.
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized, powder, white, ~13 U/mg
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Pseudomonas fluorescens, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase, recombinant, expressed in E. coli