Merck
  • Facile Synthesis of Clickable, Water-soluble and Degradable Polyphosphoesters.
Facile Synthesis of Clickable, Water-soluble and Degradable Polyphosphoesters.
ACS macro letters (2012-08-07)
Shiyi Zhang, Ang Li, Jiong Zou, Lily Yun Lin, Karen L Wooley
ABSTRACT
"Click" chemistry is a library of efficient and reliable reactions, which have been used to functionalize various classes of bio- and synthetic macromolecular systems for the incorporation of designed properties and functions. In this report, azide-alkyne Huisgen cycloaddition and thiol-yne reactions, two classical "click" chemistries, were employed to functionalize biodegradable, clickable polyphosphoester homopolymers and their water-soluble copolymers. A stable alkyne-functionalized phospholane monomer was synthesized, its organocatalyzed polymerization kinetics were evaluated, and the resulting (co)polymers were utilized to develop this facile method that provides the synthesis of clickable, water-soluble and degradable polyphosphoesters, which can be adapted for various applications.
Sigma-Aldrich
Ethyl ethylene phospholane monomer