Merck
  • Characterization of the Presence and Function of Platelet Opioid Receptors.
Characterization of the Presence and Function of Platelet Opioid Receptors.
ACS measurement science Au (2022-07-19)
Sarah M Gruba, Danielle H Francis, Audrey F Meyer, Eleni Spanolios, Jiayi He, Ben M Meyer, Donghyuk Kim, Kang Xiong-Hang, Christy L Haynes
ABSTRACT
Opioids are typically used for the treatment of pain related to disease or surgery. In the body, they enter the bloodstream and interact with a variety of immune and neurological cells that express the μ-, δ-, and κ-opioid receptors. One blood-borne cell-like body that is not well understood in the context of opioid interactions is the platelet. The platelet is a highly sensitive anucleate cell-like fragment responsible for maintaining hemostasis through shape change and the secretion of chemical messengers. This research characterizes platelet opioid receptors, how specific receptor agonists impact platelet exocytosis, and the role of the κ-and μ-receptors in platelet function. Platelets were found to express all three opioid receptors, but upon stimulation with their respective agonist no activation was detected. Furthermore, exposure to the opioid agonists did not impact traditional platelet secretion stimulated by thrombin, a natural platelet activator. In addition, data collected from knockout mice suggest that the opioid agonists may be interacting nonspecifically with platelets. Dark-field images revealed differences in activated platelet shape between the κ- and μ-knockout platelets and the control platelets. Finally, κ-knockout platelets showed variations in their ability to adhere and aggregate compared to control platelets. Overall, these data show that platelet function is not likely to be heavily affected by blood-borne opioids.
Sigma-Aldrich
Anti-Opioid Receptor Antibody, δ, pain, NT, serum, Chemicon®
Pricing and availability is not currently available.