Merck
  • TDP-43 mediates SREBF2-regulated gene expression required for oligodendrocyte myelination.
TDP-43 mediates SREBF2-regulated gene expression required for oligodendrocyte myelination.
The Journal of cell biology (2021-08-05)
Wan Yun Ho, Jer-Cherng Chang, Kenneth Lim, Amaury Cazenave-Gassiot, Aivi T Nguyen, Juat Chin Foo, Sneha Muralidharan, Ashley Viera-Ortiz, Sarah J M Ong, Jin Hui Hor, Ira Agrawal, Shawn Hoon, Olubankole Aladesuyi Arogundade, Maria J Rodriguez, Su Min Lim, Seung Hyun Kim, John Ravits, Shi-Yan Ng, Markus R Wenk, Edward B Lee, Greg Tucker-Kellogg, Shuo-Chien Ling
ABSTRACT
Cholesterol metabolism operates autonomously within the central nervous system (CNS), where the majority of cholesterol resides in myelin. We demonstrate that TDP-43, the pathological signature protein for amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD), influences cholesterol metabolism in oligodendrocytes. TDP-43 binds directly to mRNA of SREBF2, the master transcription regulator for cholesterol metabolism, and multiple mRNAs encoding proteins responsible for cholesterol biosynthesis and uptake, including HMGCR, HMGCS1, and LDLR. TDP-43 depletion leads to reduced SREBF2 and LDLR expression, and cholesterol levels in vitro and in vivo. TDP-43-mediated changes in cholesterol levels can be restored by reintroducing SREBF2 or LDLR. Additionally, cholesterol supplementation rescues demyelination caused by TDP-43 deletion. Furthermore, oligodendrocytes harboring TDP-43 pathology from FTD patients show reduced HMGCR and HMGCS1, and coaggregation of LDLR and TDP-43. Collectively, our results indicate that TDP-43 plays a role in cholesterol homeostasis in oligodendrocytes, and cholesterol dysmetabolism may be implicated in TDP-43 proteinopathies-related diseases.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Sigma-Aldrich
Cholesterol/Cholesteryl Ester Quantitation Kit