Merck
  • Synthesis and QSAR of quinazoline sulfonamides as highly potent human histamine H4 receptor inverse agonists.
Synthesis and QSAR of quinazoline sulfonamides as highly potent human histamine H4 receptor inverse agonists.
Journal of medicinal chemistry (2010-03-03)
Rogier A Smits, Maristella Adami, Enade P Istyastono, Obbe P Zuiderveld, Cindy M E van Dam, Frans J J de Kanter, Aldo Jongejan, Gabriella Coruzzi, Rob Leurs, Iwan J P de Esch
ABSTRACT
Hit optimization of the class of quinazoline containing histamine H(4) receptor (H(4)R) ligands resulted in a sulfonamide substituted analogue with high affinity for the H(4)R. This moiety leads to improved physicochemical properties and is believed to probe a distinct H(4)R binding pocket that was previously identified using pharmacophore modeling. By introducing a variety of sulfonamide substituents, the H(4)R affinity was optimized. The interaction of the new ligands, in combination with a set of previously published quinazoline compounds, was described by a QSAR equation. Pharmacological studies revealed that the sulfonamide analogues have excellent H(4)R affinity and behave as inverse agonists at the human H(4)R. In vivo evaluation of the potent 2-(6-chloro-2-(4-methylpiperazin-1-yl)quinazoline-4-amino)-N-phenylethanesulfonamide (54) (pK(i) = 8.31 +/- 0.10) revealed it to have anti-inflammatory activity in an animal model of acute inflammation.
Sigma-Aldrich
Histamine, ≥97.0%
Pricing and availability is not currently available.