Merck
  • Structural Dependence of Reductive Defluorination of Linear PFAS Compounds in a UV/Electrochemical System.
Structural Dependence of Reductive Defluorination of Linear PFAS Compounds in a UV/Electrochemical System.
Environmental science & technology (2020-08-14)
Unnati Rao, Yiming Su, Chia Miang Khor, Bongyeon Jung, Shengcun Ma, David M Cwiertny, Bryan M Wong, David Jassby
ABSTRACT
Per and polyfluoroalkyl substances (PFAS), legacy chemicals used in firefighting and the manufacturing of many industrial and consumer goods, are widely found in groundwater resources, along with other regulated compounds, such as chlorinated solvents. Due to their strong C-F bonds, these molecules are extremely recalcitrant, requiring advanced treatment methods for effective remediation, with hydrated electrons shown to be able to defluorinated these compounds. A combined photo/electrochemical method has been demonstrated to dramatically increase defluorination rates, where PFAS molecules sorbed onto appropriately functionalized cathodes charged to low cell potentials (-0.58 V vs Ag/AgCl) undergo a transient electron transfer event from the electrode, which "primes" the molecule by reducing the C-F bond strength and enables the bond's dissociation upon the absorption of a hydrated electron. In this work, we explore the impact of headgroup and chain length on the performance of this two-electron process and extend this technique to chlorinated solvents. We use isotopically labeled PFAS molecules to take advantage of the kinetic isotope effect and demonstrate that indeed PFAS defluorination is likely driven by a two-electron process. We also present density functional theory calculations to illustrate that the externally applied potential resulted in an increased rate of electron transfer, which ultimately increased the measured defluorination rate.
Sigma-Aldrich
Nonafluorobutane-1-sulfonic acid, 97%
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%