Merck
  • Stage-dependent Increase in Expression of miR-155 and Ki-67 and Number of Tumour-associated Inflammatory Cells in Folliculotropic Mycosis Fungoides.
Stage-dependent Increase in Expression of miR-155 and Ki-67 and Number of Tumour-associated Inflammatory Cells in Folliculotropic Mycosis Fungoides.
Acta dermato-venereologica (2020-06-20)
Lihi Atzmony, Lilach Moyal, Meora Feinmesser, Batya Gorovitz, Avraham Hirshberg, Iris Amitay-Laish, Hadas Prag-Naveh, Aviv Barzilai, Emmilia Hodak
ABSTRACT
Recent studies suggest that folliculotropic mycosis fungoides (FMF), the most common variant of mycosis fungoides (MF), presents with 2 distinct clinicopathological stages: early indolent stage and more aggressive advanced/tumour stage. To further characterize these stages, miR-155 expression was studied with qRT-PCR and found to be significantly higher in biopsies of tumour-stage FMF compared with early-stage FMF and inflammatory dermatoses. There was no statistically significant difference in miR-155 expression between early-stage FMF and early-stage MF, nor between tumour-stage FMF and tumour-stage MF. Immunohistochemical analysis revealed a significantly increased number of dermal Ki-67+ proliferating lymphocytes in tumour-stage FMF, together with an increased number of CD20+ B cells and CD68+ macrophages compared with early-stage FMF. Thus, similar to classic MF, miR-155, Ki-67 tumour cell immunoreactivity, and certain tumour-infiltrating inflammatory cells are differentially expressed in early- vs tumour-stage FMF. The results of this study corroborate the notion that FMF presents with 2 distinct stages.
Sigma-Aldrich
Anti-TOX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution