- Detection of Escherichia coli O157:H7 Using Automated Immunomagnetic Separation and Enzyme-Based Colorimetric Assay.
Detection of Escherichia coli O157:H7 Using Automated Immunomagnetic Separation and Enzyme-Based Colorimetric Assay.
The food industry requires rapid and simple detection methods for preventing harm from pathogenic bacteria. Until now, various technologies used to detect foodborne bacteria were time-consuming and laborious. Therefore, we have developed an automated immunomagnetic separation combined with a colorimetric assay for the rapid detection of E. coli O157:H7 in food samples. The colorimetric detection method using enzymatic reaction is fascinating because of its simplicity and rapidity and does not need sophisticated devices. Moreover, the proposed procedures for the detection of bacteria in food take less than 3 h including pre-enrichment, separation and detection steps. First, target-specific immunomagnetic beads were introduced to contaminated milk in a pre-enrichment step. Second, the pre-enriched sample solution containing target bacteria bound on immunomagnetic beads was injected into an automated pretreatment system. Subsequently, the immunomagnetic beads along with target bacteria were separated and concentrated into a recovery tube. Finally, released β-galactosidase from E. coli O157:H7 after lysis was reacted with chlorophenol red β-galactopyranoside (CPRG) used as a substrate and the colorimetric change of CPRG was determined by absorbance measuring or the naked eye. By the proposed approach in this study, we could detect 3 × 102 CFU/mL of E. coli O157:H7 from a milk sample within 3 h.