MilliporeSigma
  • Discovery of a novel class of covalent inhibitor for aldehyde dehydrogenases.
Discovery of a novel class of covalent inhibitor for aldehyde dehydrogenases.
The Journal of biological chemistry (2011-10-25)
May Khanna, Che-Hong Chen, Ann Kimble-Hill, Bibek Parajuli, Samantha Perez-Miller, Sulochanadevi Baskaran, Jeewon Kim, Karl Dria, Vasilis Vasiliou, Daria Mochly-Rosen, Thomas D Hurley
ABSTRACT
Human aldehyde dehydrogenases (ALDHs) comprise a family of 17 homologous enzymes that metabolize different biogenic and exogenic aldehydes. To date, there are relatively few general ALDH inhibitors that can be used to probe the contribution of this class of enzymes to particular metabolic pathways. Here, we report the discovery of a general class of ALDH inhibitors with a common mechanism of action. The combined data from kinetic studies, mass spectrometric measurements, and crystallographic analyses demonstrate that these inhibitors undergo an enzyme-mediated β-elimination reaction generating a vinyl ketone intermediate that covalently modifies the active site cysteine residue present in these enzymes. The studies described here can provide the basis for rational approach to design ALDH isoenzyme-specific inhibitors as research tools and perhaps as drugs, to address diseases such as cancer where increased ALDH activity is associated with a cellular phenotype.
Sigma-Aldrich
MTT Cell Growth Assay Kit, MTT Cell Growth Assay is a colorimetric assay that can be used for either proliferation or complement-mediated cytotoxicity assays.