Merck
  • Regioselective Single-Electron Tsuji-Trost Reaction of Allylic Alcohols: A Photoredox/Nickel Dual Catalytic Approach.
Regioselective Single-Electron Tsuji-Trost Reaction of Allylic Alcohols: A Photoredox/Nickel Dual Catalytic Approach.
Organic letters (2019-08-08)
Zheng-Jun Wang, Shuai Zheng, Eugénie Romero, Jennifer K Matsui, Gary A Molander
ABSTRACT
A radical-mediated functionalization of allyl alcohol derived partners with a variety of alkyl 1,4-dihydropyridines via photoredox/nickel dual catalysis is described. This transformation transpires with high linear and E-selectivity, avoiding the requirement of harsh conditions (e.g., strong base, elevated temperature). Additionally, using aryl sulfinate salts as radical precursors, allyl sulfones can also be obtained. Kinetic isotope effect experiments implicated oxidative addition of the nickel catalyst to the allylic electrophile as the turnover-limiting step, supporting previous computational studies.