Merck
  • Molecular basis for metabolite channeling in a ring opening enzyme of the phenylacetate degradation pathway.
Molecular basis for metabolite channeling in a ring opening enzyme of the phenylacetate degradation pathway.
Nature communications (2019-09-13)
Nitish Sathyanarayanan, Giuseppe Cannone, Lokesh Gakhar, Nainesh Katagihallimath, Ramanathan Sowdhamini, Subramanian Ramaswamy, Kutti R Vinothkumar
ABSTRACT
Substrate channeling is a mechanism for the internal transfer of hydrophobic, unstable or toxic intermediates from the active site of one enzyme to another. Such transfer has previously been described to be mediated by a hydrophobic tunnel, the use of electrostatic highways or pivoting and by conformational changes. The enzyme PaaZ is used by many bacteria to degrade environmental pollutants. PaaZ is a bifunctional enzyme that catalyzes the ring opening of oxepin-CoA and converts it to 3-oxo-5,6-dehydrosuberyl-CoA. Here we report the structures of PaaZ determined by electron cryomicroscopy with and without bound ligands. The structures reveal that three domain-swapped dimers of the enzyme form a trilobed structure. A combination of small-angle X-ray scattering (SAXS), computational studies, mutagenesis and microbial growth experiments suggests that the key intermediate is transferred from one active site to the other by a mechanism of electrostatic pivoting of the CoA moiety, mediated by a set of conserved positively charged residues.
Sigma-Aldrich
Graphene oxide, 2 mg/mL, dispersion in H2O, avg. no. of layers, 1
Pricing and availability is not currently available.