Merck
  • Isoflavones prevent oxidative stress and inhibit the activity of the enzyme monoamine oxidase in vitro.
Isoflavones prevent oxidative stress and inhibit the activity of the enzyme monoamine oxidase in vitro.
Molecular biology reports (2019-02-14)
Izaviany da Silva Schmitz, Larissa Finger Schaffer, Alcindo Busanello, Catiuscia Molz de Freitas, Roselei Fachinetto, Luis Ricardo Peroza
ABSTRACT
Oxidative stress occurs due to an imbalance between antioxidant defenses and pro-oxidant agents in brain. This condition has been associated to the pathogenesis of several brain diseases; therefore, increasing the use of compounds that exert antioxidant activity. Thus, the objective of this study was to evaluate, in vitro, the effect of isoflavones in: (1) lipid peroxidation, catalase activity and thiol groups in the presence of pro-oxidants: sodium nitroprusside or Fe2+/EDTA complex in rat brain homogenates; (2) the activity of the enzyme monoamine oxidase (MAO). As a result, the isoflavones reduced lipid peroxidation in a manner dependent on the concentration and protected against the reduction of catalase activity as well as the induced thiol oxidation in brain tissue. In addition, isoflavones inhibited MAO activity (MAO-A and MAO-B). Taken together, our results showed that isoflavones avoided oxidative stress and decreased the MAO activity, suggesting a promissory use in the treatment of neurodegenerative diseases.