Merck
  • High-throughput production of gene replacement mutants in Neurospora crassa.
High-throughput production of gene replacement mutants in Neurospora crassa.
Methods in molecular biology (Clifton, N.J.) (2011-05-19)
Gyungsoon Park, Hildur V Colot, Patrick D Collopy, Svetlana Krystofova, Christopher Crew, Carol Ringelberg, Liubov Litvinkova, Lorena Altamirano, Liande Li, Susan Curilla, Wei Wang, Norma Gorrochotegui-Escalante, Jay C Dunlap, Katherine A Borkovich
ABSTRACT
The model filamentous fungus Neurospora crassa has been the focus of functional genomics studies for the past several years. A high-throughput gene knockout procedure has been developed and used to generate mutants for more than two-thirds of the ∼10,000 annotated N. crassa genes. Yeast recombinational cloning was incorporated as an efficient procedure to produce all knockout cassettes. N. crassa strains with the Δmus-51 or Δmus-52 deletion mutations were used as transformation recipients in order to reduce the incidence of ectopic integration and increase homologous recombination of knockout cassettes into the genome. A 96-well format was used for many steps of the procedure, including fungal transformation, isolation of homokaryons, and verification of mutants. In addition, development of software programs for primer design and restriction enzyme selection facilitated the high-throughput aspects of the overall protocol.
Sigma-Aldrich
Fibrin In Vitro Angiogenesis Assay, The Fibrin Gel In Vitro Angiogenesis Assay Kit represents a simple model of angiogenesis in which the induction or inhibition of tube formation by exogenous signals can be easily monitored.
Pricing and availability is not currently available.