
DMT-2′Fluoro-dC(ac) Phosphoramidite

Synonym(s):
DMT-2′-F-dC(ac) amidite
Empirical Formula (Hill Notation):
C41H49FN5O8P
CAS Number:
Molecular Weight:
789.83
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing