Skip to Content
Merck
All Photos(1)

Key Documents

PZ0343

Sigma-Aldrich

PF-06260414

≥98% (HPLC)

Synonym(s):

6-[(4R)-tetrahydro-4-methyl-1,1-dioxido-2H-1,2,6-thiadiazin-2-yl]-1-isoquinolinecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O2S
CAS Number:
Molecular Weight:
302.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

C[C@H](CNS1(=O)=O)CN1C2=CC3=CC=NC(C#N)=C3C=C2

InChI

1S/C14H14N4O2S/c1-10-8-17-21(19,20)18(9-10)12-2-3-13-11(6-12)4-5-16-14(13)7-15/h2-6,10,17H,8-9H2,1H3/t10-/m1/s1

InChI key

ZKAVFOXYJCREBQ-SNVBAGLBSA-N

Biochem/physiol Actions

PF-06260414 is a nonsteriodal selective androgen receptor modulator (SARM). PF-06260414 is being investigated as a treatment for muscle weakening such as that associated with muscular dystrophy and the sarcopenia associated with natural aging. PF-06260414 has anabolic activity without unwanted androgenic activity on reproductive organs, liver or kidneys.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emiliano Ventura et al.
Journal of chromatography. A, 1600, 183-196 (2019-05-06)
A semi-quantitative method was developed to monitor the misuse of 15 SARM compounds belonging to nine different families, in urine matrices from a range of species (equine, canine, human, bovine and murine). SARM residues were extracted from urine (200 μL) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service