MilliporeSigma
GC Analysis of Menthol Enantiomers on α-DEX 120
GC Analysis of Menthol Enantiomers on α-DEX™ 120 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
α-DEX 120
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
α-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24310)
oven
110 °C
detector
FID, 300 °C
carrier gas
helium, 20 cm/sec
injection
1 μL, 100:1 split
sample
0.5 mg/mL each component
Description
Legal Information
DEX is a trademark of Sigma-Aldrich Co. LLC