MilliporeSigma
  • Unraveling vasotocinergic, isotocinergic and stress pathways after food deprivation and high stocking density in the gilthead sea bream.
Unraveling vasotocinergic, isotocinergic and stress pathways after food deprivation and high stocking density in the gilthead sea bream.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2017-10-29)
Arleta Krystyna Skrzynska, Juan Antonio Martos-Sitcha, Gonzalo Martínez-Rodríguez, Juan Miguel Mancera
ABSTRACT
The influence of chronic stress, induced by food deprivation (FD) and/or high stocking density (HSD), was assessed on stress, vasotocinergic and isotocinergic pathways of the gilthead sea bream (Sparus aurata). Fish were randomly assigned to one of the following treatments: (1) fed at low stocking density (LSD-F; 5kg·m
Sigma-Aldrich
Heparin ammonium salt from porcine intestinal mucosa, ≥140 USP units/mg