MilliporeSigma
  • Silence of inositol 1,4,5-trisphosphate receptor expression decreases cyantraniliprole susceptibility in Bemisia tabaci.
Silence of inositol 1,4,5-trisphosphate receptor expression decreases cyantraniliprole susceptibility in Bemisia tabaci.
Pesticide biochemistry and physiology (2017-11-07)
Lei Guo, Pei Liang, Kuan Fang, Dong Chu
ABSTRACT
Cyantraniliprole is the second active ingredient of anthranilic diamide insecticide, and the first to control a cross-spectrum of chewing and sucking pests such as sweetpotato whitefly, Bemisia tabaci (Gennadius) (Hemiptera: Aleyrodidae). The inositol 1,4,5-trisphosphate receptor (IP3R) and ryanodine receptor (RyR) are two families of Ca
Product Description
Supelco
Cyantraniliprole, PESTANAL®, analytical standard