MilliporeSigma
Molecular Serotype-Specific Identification of Non-type b
Frontiers in microbiology (2017-10-20)
Chika Takano, Mitsuko Seki, Dong Wook Kim, Paul E Kilgore, Kazumasa Fuwa, Koji Takahashi, Toshiaki Inazaki, Satoshi Hayakawa
ABSTRACT
Over the past four decades, the incidence of meningitis caused by
Sigma-Aldrich
Triphenylmethane, 99%