MilliporeSigma
  • Interleukin-6 Deficiency Attenuates Retinal Ganglion Cell Axonopathy and Glaucoma-Related Vision Loss.
Interleukin-6 Deficiency Attenuates Retinal Ganglion Cell Axonopathy and Glaucoma-Related Vision Loss.
Frontiers in neuroscience (2017-06-18)
Franklin D Echevarria, Cathryn R Formichella, Rebecca M Sappington
ABSTRACT
The pleotropic cytokine interleukin-6 (IL-6) is implicated in retinal ganglion cell (RGC) survival and degeneration, including that associated with glaucoma. IL-6 protects RGCs from pressure-induced apoptosis
Sigma-Aldrich
CTB, ≥98% (HPLC)