MilliporeSigma
  • Determination of absolute configuration using kinetic resolution catalysts.
Determination of absolute configuration using kinetic resolution catalysts.
Organic letters (2011-07-23)
Alexander J Wagner, Jonathan G David, Scott D Rychnovsky
ABSTRACT
A new method was developed to assign the absolute configuration of molecules using kinetic resolution catalysts. Secondary alcohols were acylated in the presence of Birman's S-HBTM and R-HBTM catalysts, and the fast-reacting catalyst was identified by NMR analysis of the reaction mixture. A mnemonic was developed to assign configuration based on the identity of the fast-reacting catalyst. The method uses only 1-3 mg of alcohol, and it is more convenient than the Mosher method. The kinetic resolution strategy may be extended to other classes of molecules.
Sigma-Aldrich
(2R)-2-Phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole, ≥95%