MilliporeSigma
  • 5-HTR3 and 5-HTR4 located on the mitochondrial membrane and functionally regulated mitochondrial functions.
5-HTR3 and 5-HTR4 located on the mitochondrial membrane and functionally regulated mitochondrial functions.
Scientific reports (2016-11-23)
Qingyi Wang, Huiyuan Zhang, Hao Xu, Dongqing Guo, Hui Shi, Yuan Li, Weiwei Zhang, Yuchun Gu
ABSTRACT
5-HT has been reported to possess significant effects on cardiac activities, but activation of 5-HTR on the cell membrane failed to illustrate the controversial cardiac reaction. Because 5-HT constantly comes across the cell membrane via 5-HT transporter (5-HTT) into the cytoplasm, whether 5-HTR is functional present on the cellular organelles is unknown. Here we show 5-HTR3 and 5-HTR4 were located in cardiac mitochondria, and regulated mitochondrial activities and cellular functions. Knock down 5-HTR3 and 5-HTR4 in neonatal cardiomyocytes resulted in significant increase of cell damage in response to hypoxia, and also led to alternation in heart beating. Activation of 5-HTR4 attenuated mitochondrial Ca
Sigma-Aldrich
RS 23597-190, ≥98% (HPLC)