MilliporeSigma
  • Role of the Flt-1 receptor tyrosine kinase in regulating the assembly of vascular endothelium.
Role of the Flt-1 receptor tyrosine kinase in regulating the assembly of vascular endothelium.
Nature (1995-07-06)
G H Fong, J Rossant, M Gertsenstein, M L Breitman
ABSTRACT
The vascular endothelial growth factor (VEGF) and its high-affinity binding receptors, the tyrosine kinases Flt-1 and Flk-1, are thought to be important for the development of embryonic vasculature. Here we report that Flt-1 is essential for the organization of embryonic vasculature, but is not essential for endothelial cell differentiation. Mouse embryos homozygous for a targeted mutation in the flt-1 locus, flt-1lcz, formed endothelial cells in both embryonic and extra-embryonic regions, but assembled these cells into abnormal vascular channels and died in utero at mid-somite stages. At earlier stages, the blood islands of flt-1lcz homozygotes were abnormal, with angioblasts in the interior as well as on the periphery. We suggest that the Flt-1 signalling pathway may regulate normal endothelial cell-cell or cell-matrix interactions during vascular development.
Sigma-Aldrich
FLT1 (784-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution