MilliporeSigma
  • Methyl jasmonate elicits the biotransformation of geraniol stored as its glucose conjugate into methyl geranate in Achyranthes bidentata plant.
Methyl jasmonate elicits the biotransformation of geraniol stored as its glucose conjugate into methyl geranate in Achyranthes bidentata plant.
Plant physiology and biochemistry : PPB (2016-10-04)
Shigeru Tamogami, Ganesh K Agrawal, Randeep Rakwal
ABSTRACT
To investigate the biotransformation pathway of airborne geraniol by Achyranthes bidentata (A. bidentata), deuterium labeled geraniol was applied with or without methyl jasmonate (MeJA), and the biosynthesized metabolites were analyzed. In A. bidentata leaves, geraniol was conjugated with glucose. The conjugate was then metabolized to afford methyl geranate only under MeJA elicitation. MeJA elicits the biotransformation of geraniol into methyl geranate by inducing the conversion of the intermediate, glucose conjugate of geraniol.
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil