MilliporeSigma
  • Human recombinant antibodies against Plasmodium falciparum merozoite surface protein 3 cloned from peripheral blood leukocytes of individuals with immunity to malaria demonstrate antiparasitic properties.
Human recombinant antibodies against Plasmodium falciparum merozoite surface protein 3 cloned from peripheral blood leukocytes of individuals with immunity to malaria demonstrate antiparasitic properties.
Infection and immunity (2006-05-23)
Rasmus Lundquist, Leif Kofoed Nielsen, Ali Jafarshad, Daw Soesoe, Lars Harder Christensen, Pierre Druilhe, Morten Hanefeld Dziegiel
ABSTRACT
Immunoglobulins from individuals with immunity to malaria have a strong antiparasitic effect when transferred to Plasmodium falciparum malaria infected patients. One prominent target of antiparasitic antibodies is the merozoite surface antigen 3 (MSP-3). We have investigated the antibody response against MSP-3 residues 194 to 257 (MSP-3(194-257)) on the molecular level. mRNA from peripheral blood leukocytes from clinically immune individuals was used as a source of Fab (fragment antibody) genes. A Fab-phage display library was made, and three distinct antibodies designated RAM1, RAM2, and RAM3 were isolated by panning. Immunoglobulin G1 (IgG1) and IgG3 full-length antibodies have been produced in CHO cells. Reactivity with the native parasite protein was demonstrated by immunofluorescence microscopy, flow cytometry, and immunoblotting. Furthermore, the antiparasitic effect of RAM1 has been tested in vitro in an antibody-dependent cellular inhibition (ADCI) assay. Both the IgG1 and the IgG3 versions of the antibody show an inhibitory effect on parasite growth.
Product Description
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O