MilliporeSigma
  • Identification and characterization of a novel class of interleukin-1 post-translational processing inhibitors.
Identification and characterization of a novel class of interleukin-1 post-translational processing inhibitors.
The Journal of pharmacology and experimental therapeutics (2001-09-19)
D G Perregaux, P McNiff, R Laliberte, N Hawryluk, H Peurano, E Stam, J Eggler, R Griffiths, M A Dombroski, C A Gabel
ABSTRACT
Lipopolysaccharide (LPS)-activated monocytes and macrophages produce large quantities of pro-interleukin (IL)-1beta but externalize little mature cytokine. Efficient post-translational processing of the procytokine occurs in vitro when these cells encounter a secretion stimulus such as ATP, cytolytic T cells, or hypotonic stress. Each of these stimuli promotes rapid conversion of 31-kDa pro-IL-1beta to its mature 17-kDa species and release of the 17-kDa cytokine. In this study, two novel pharmacological agents, CP-424,174 and CP-412,245, are identified as potent inhibitors of stimulus-coupled IL-1beta post-translational processing. These agents, both diarylsulfonylureas, block formation of mature IL-1beta without increasing the amount of procytokine that is released extracellularly, and they inhibit independently of the secretion stimulus used. Conditioned medium derived from LPS-activated/ATP-treated human monocytes maintained in the absence and presence of CP-424,174 contained comparable quantities of IL-6, tumor necrosis factor-alpha (TNFalpha), and IL-1RA, but 30-fold less IL-1beta was generated in the test agent's presence. As a result of this decrease, monocyte conditioned medium prepared in the presence of CP-424,174 demonstrated a greatly diminished capacity to promote an IL-1-dependent response (induction of serum amyloid A synthesis by Hep3B cells). Oral administration of CP-424,174 to mice resulted in inhibition of IL-1 in the absence of an effect on IL-6 and TNFalpha. These novel agents, therefore, act as selective cytokine release inhibitors and define a new therapeutic approach for controlling IL-1 production in inflammatory diseases.
Sigma-Aldrich
CP-424,174, ≥98% (HPLC)