MilliporeSigma
  • Time-dependent activity of Na+/H+ exchanger isoform 1 and homeostasis of intracellular pH in astrocytes exposed to CoCl2 treatment.
Time-dependent activity of Na+/H+ exchanger isoform 1 and homeostasis of intracellular pH in astrocytes exposed to CoCl2 treatment.
Molecular medicine reports (2016-04-02)
Peng Wang, Ling Li, Zhenxiang Zhang, Quancheng Kan, Feng Gao, Suyan Chen
ABSTRACT
Hypoxia causes injury to the central nervous system during stroke and has significant effects on pH homeostasis. Na+/H+ exchanger isoform 1 (NHE1) is important in the mechanisms of hypoxia and intracellular pH (pHi) homeostasis. As a well-established hypoxia-mimetic agent, CoCl2 stabilizes and increases the expression of hypoxia inducible factor‑1α (HIF-1α), which regulates several genes involved in pH balance, including NHE1. However, it is not fully understood whether NHE1 is activated in astrocytes under CoCl2 treatment. In the current study, pHi and NHE activity were analyzed using the pHi‑sensitive dye BCECF‑AM. Using cariporide (an NHE1‑specific inhibitor) and EIPA (an NHE nonspecific inhibitor), the current study demonstrated that it was NHE1, not the other NHE isoforms, that was important in regulating pHi homeostasis in astrocytes during CoCl2 treatment. Additionally, the present study observed that, during the early period of CoCl2 treatment (the first 2 h), NHE1 activity and pHi dropped immediately, and NHE1 mRNA expression was reduced compared with control levels, whereas expression levels of the NHE1 protein had not yet changed. In the later period of CoCl2 treatment, NHE1 activity and pHi significantly increased compared with the control levels, as did the mRNA and protein expression levels of NHE1. Furthermore, the cell viability and injury of astrocytes was not changed during the initial 8 h of CoCl2 treatment; their deterioration was associated with the higher levels of pHi and NHE1 activity. The current study concluded that NHE1 activity and pHi homeostasis are regulated by CoCl2 treatment in a time-dependent manner in astrocytes, and may be responsible for the changes in cell viability and injury observed under hypoxia-mimetic conditions induced by CoCl2 treatment.
Sigma-Aldrich
Anti-GFAP antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP78, affinity isolated antibody