MilliporeSigma
  • Regulation of cysteine cathepsin expression by oxidative stress in the retinal pigment epithelium/choroid of the mouse.
Regulation of cysteine cathepsin expression by oxidative stress in the retinal pigment epithelium/choroid of the mouse.
Experimental eye research (2006-05-11)
Parvaneh Alizadeh, Zeljka Smit-McBride, Sharon L Oltjen, Leonard M Hjelmeland
ABSTRACT
Cystatin C is the major inhibitor of the cysteine cathepsins. Polymorphisms in the cystatin C gene have recently been associated with the risk of developing Age-related Macular Degeneration (AMD). Oxidative stress is also thought to play a key role in the pathogenesis of AMD. We surveyed the retinal pigment epithelium (RPE) and choroid of the C57BL/6J mouse for the expression of the cysteine cathepsins under normoxic and hyperoxic (75% O(2)) conditions. Microarray analysis of RPE/choroid mRNA revealed the expression of cathepsins B and L, as well as cystatin C under all experimental conditions. The microarray results were confirmed by real-time quantitative polymerase chain reaction (PCR). Localization of the mRNA species for cystatin C and cathepsin B, as well as, localization of protein species for cystatin C, cathepsins B and L were performed to evaluate the tissue distribution of these species. Our results indicate that cystatin C is largely synthesized in the RPE and secreted from the basal side. Cathepsin B is the major cysteine protease in the RPE and choroid. The expression of all mRNAs and proteins was elevated by exposure to oxidative stress.
Product Description
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting