MilliporeSigma
  • Patient-derived bladder cancer xenografts in the preclinical development of novel targeted therapies.
Patient-derived bladder cancer xenografts in the preclinical development of novel targeted therapies.
Oncotarget (2015-06-05)
Wolfgang Jäger, Hui Xue, Tetsutaro Hayashi, Claudia Janssen, Shannon Awrey, Alexander W Wyatt, Shawn Anderson, Igor Moskalev, Anne Haegert, Mohammed Alshalalfa, Nicholas Erho, Elai Davicioni, Ladan Fazli, Estelle Li, Colin Collins, Yuzhuo Wang, Peter C Black
ABSTRACT
Optimal animal models of muscle invasive bladder cancer (MIBC) are necessary to overcome the current lack of novel targeted therapies for this malignancy. Here we report on the establishment and characterization of patient-derived primary xenografts (PDX). Patient tumors were grafted under the renal capsule of mice and subsequently transplanted over multiple generations. Patient tumor and PDX were processed for analysis of copy number variations by aCGH, gene expression by microarray, and expression of target pathways by immunohistochemistry (IHC). One PDX harbouring an FGFR3 mutation was treated with an inhibitory monoclonal antibody targeting FGFR3. Five PDX were successfully established. Tumor doubling time ranged from 5 to 11 days. Array CGH revealed shared chromosomal aberrations in the patient tumors and PDX. Gene expression microarray and IHC confirmed that PDXs maintain similar patterns to the parental tumors. Tumor growth in the PDX with an FGFR3 mutation was inhibited by the FGFR3 inhibitor. PDXs recapitulate the tumor biology of the patients' primary tumors from which they are derived. Investigations related to tumor biology and drug testing in these models are therefore more likely to be relevant to the disease state in patients. They represent a valuable tool for developing precision therapy in MIBC.
Sigma-Aldrich
Anti-Fibroblast Growth Factor Receptor-3, Cytoplasmic antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse