MilliporeSigma
  • The neuron-specific protein PGP 9.5 is a ubiquitin carboxyl-terminal hydrolase.
The neuron-specific protein PGP 9.5 is a ubiquitin carboxyl-terminal hydrolase.
Science (New York, N.Y.) (1989-11-03)
K D Wilkinson, K M Lee, S Deshpande, P Duerksen-Hughes, J M Boss, J Pohl
ABSTRACT
A complementary DNA (cDNA) for ubiquitin carboxyl-terminal hydrolase isozyme L3 was cloned from human B cells. The cDNA encodes a protein of 230 amino acids with a molecular mass of 26.182 daltons. The human protein is very similar to the bovine homolog, with only three amino acids differing in over 100 residues compared. The amino acid sequence deduced from the cDNA was 54% identical to that of the neuron-specific protein PGP 9.5. Purification of bovine PGP 9.5 confirmed that it is also a ubiquitin carboxyl-terminal hydrolase. These results suggest that a family of such related proteins exists and that their expression is tissue-specific.