MilliporeSigma
GRK6 expression in patients with hepatocellular carcinoma.
Asian Pacific journal of tropical medicine (2013-02-05)
Ya-Ping Li
ABSTRACT
To investigate the expression and potential roles of G protein-coupled receptor kinase 6 (GRK6) in hepatocellular carcinoma (HCC) patients. Immunohistochemistry and Western blot was performed to determine GRK6 expression in 73 HCC samples. And the correlation with clinicopathological features was also analyzed. GRK6 expression was significantly higher in HCC than that in normal hepatic tissue. GRK6 was positively correlated with proliferation marker Ki-67, clinical stage, metastasis and survival time. Our results suggested that GRK6 overexpression plays an important role in HCC. Monitoring the expression of GRK6 maybe helpful in early diagnosis and prognosis of HCC.