MilliporeSigma
  • TMEM126A, encoding a mitochondrial protein, is mutated in autosomal-recessive nonsyndromic optic atrophy.
TMEM126A, encoding a mitochondrial protein, is mutated in autosomal-recessive nonsyndromic optic atrophy.
American journal of human genetics (2009-03-31)
Sylvain Hanein, Isabelle Perrault, Olivier Roche, Sylvie Gerber, Noman Khadom, Marlene Rio, Nathalie Boddaert, Marc Jean-Pierre, Nora Brahimi, Valérie Serre, Dominique Chretien, Nathalie Delphin, Lucas Fares-Taie, Sahran Lachheb, Agnès Rotig, Françoise Meire, Arnold Munnich, Jean-Louis Dufier, Josseline Kaplan, Jean-Michel Rozet
ABSTRACT
Nonsyndromic autosomal-recessive optic neuropathies are rare conditions of unknown genetic and molecular origin. Using an approach of whole-genome homozygosity mapping and positional cloning, we have identified the first gene, to our knowledge, responsible for this condition, TMEM126A, in a large multiplex inbred Algerian family and subsequently in three other families originating from the Maghreb. TMEM126A is conserved in higher eukaryotes and encodes a transmembrane mitochondrial protein of unknown function, supporting the view that mitochondrial dysfunction may be a hallmark of inherited optic neuropathies including isolated autosomal-recessive forms.